Learn More
MCTP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530761
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: U87-MG. Predicted reactivity: Mouse (97%), Rat (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Might play a role in the development of cardiac outflow tract. [UniProt]Specifications
MCTP2 | |
Polyclonal | |
Unconjugated | |
MCTP2 | |
Gm489; MCTP2; multiple C2 and transmembrane domain containing 2; multiple C2 and transmembrane domain-containing protein 2; multiple C2 domains, transmembrane 2; multiple C2-domains with two transmembrane regions 2 1; RGD1562967 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55784 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.51 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q6DN12 | |
MCTP2 | |
Recombinant fragment corresponding to a region within amino acids 471 and 693 of Human MCTP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |