missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCT5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568230
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
MCT5 (SLC16A4, Solute Carrier Family 16 Member 4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include symporter activity and monocarboxylic acid transmembrane transporter activity. Proton-linked monocarboxylate transporter. Catalyzes the rapid transport across the plasma membrane of many monocarboxylates such as lactate, pyruvate, branched-chain oxo acids derived from leucine, valine and isoleucine, and the ketone bodies acetoacetate, beta-hydroxybutyrate and acetate.
Specifications
| MCT5 | |
| Polyclonal | |
| Unconjugated | |
| SLC16A5 | |
| A130015N09Rik; MCT 5; MCT 6; MCT5; MCT6; monocarboxylate transporter; monocarboxylate transporter 5; Monocarboxylate transporter 6; SLC16A5; solute carrier family 16 (monocarboxylate transporter), member 5; solute carrier family 16 (monocarboxylic acid transporters), member 5; solute carrier family 16 member 5; solute carrier family 16, member 5 (monocarboxylic acid transporter 6) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 217316, 690212, 9121 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| G5E8K6, O15375 | |
| SLC16A5 | |
| Synthesized peptide derived from human MCT5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction