missing translation for 'onlineSavingsMsg'
Learn More

MCT4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Product Code. 16882085 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16882085 20 μL 20µL
16872085 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16882085 Supplier Proteintech Supplier No. 227871AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Minus end-directed microtubule-dependent motor required for bipolar spindle formation. May contribute to movement of early endocytic vesicles. Regulates cilium formation and structure.

Specifications

Antigen MCT4
Applications Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry, Immunofluorescence
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLC16A3
Gene Accession No. O15427, P57787
Gene Alias MCT 4, MCT4, Monocarboxylate transporter 3, Monocarboxylate transporter 4, SLC16A3
Gene Symbols SLC16A3
Host Species Rabbit
Immunogen MCT4 Fusion Protein Ag18788
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80879, 9123
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.