missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Minus end-directed microtubule-dependent motor required for bipolar spindle formation. May contribute to movement of early endocytic vesicles. Regulates cilium formation and structure.
Specifications
Specifications
| Antigen | MCT4 |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.38 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SLC16A3 |
| Gene Accession No. | O15427, P57787 |
| Gene Alias | MCT 4, MCT4, Monocarboxylate transporter 3, Monocarboxylate transporter 4, SLC16A3 |
| Gene Symbols | SLC16A3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?