missing translation for 'onlineSavingsMsg'
Learn More

MCT3/SLC16A8 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18393862 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393862 100 μg 100µg
18301872 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18393862 Supplier Novus Biologicals Supplier No. NBP311919100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MCT3/SLC16A8 Polyclonal antibody specifically detects MCT3/SLC16A8 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCT3/SLC16A8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:400
Formulation PBS
Gene Alias MCT3MCT 3, monocarboxylate transporter 3, REMP, solute carrier 16 (monocarboxylic acid transporters), member 8, Solute carrier family 16 member 8, solute carrier family 16, member 8 (monocarboxylic acid transporter 3)
Host Species Rabbit
Immunogen Synthetic peptide derived from MCT3/SLC16A8 protein. (Uniprot: O95907)
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23539
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.