missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCT2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA577498
This item is not returnable.
View return policy
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt. Reconstitute using 50 uL deionized water.
Tissues with few or no mitochondria, such as erythrocytes and tumor cells, depend largely on glycolysis to generate ATP. The major end products of glycolysis, pyruvate and lactate, must be eliminated from these cells to enable continued glycolytic flux and prevent toxic effects. H+/monocarboxylate transporters mediate the transport of lactate and pyruvate. Human MCT2 has a high affinity for the transport of pyruvate.
Specifications
| MCT2 | |
| Polyclonal | |
| Unconjugated | |
| Slc16a7 | |
| Monocarboxylate Transporter 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20503, 29735 | |
| -20°C | |
| Lyophilized |
| Immunofluorescence, Immunohistochemistry, Western Blot | |
| 0.8 mg/mL | |
| PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
| O70451, Q63344 | |
| Slc16a7 | |
| Peptide (C)KEMEALSRSKQDD, corresponding to amino acid residues 456-468 of rat Monocarboxylate Transporter 2 | |
| 50 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction