missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MCT2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA576603

Product Code. 15953125

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Tissues with few or no mitochondria, such as erythrocytes and tumor cells, depend largely on glycolysis to generate ATP. The major end products of glycolysis, pyruvate and lactate, must be eliminated from these cells to enable continued glycolytic flux and prevent toxic effects. H+/monocarboxylate transporters mediate the transport of lactate and pyruvate. Human MCT2 has a high affinity for the transport of pyruvate.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MCT2
Polyclonal
Unconjugated
SLC16A7
4921534N07Rik; MCT 2; Mct2; monocarboxylate transporter; Monocarboxylate transporter 2; Slc16a7; solute carrier family 16 (monocarboxylate transporter), member 7; solute carrier family 16 (monocarboxylic acid transporters), member 7; solute carrier family 16 member 7; solute carrier family 16, member 7 (monocarboxylic acid transporter 2)
Rabbit
Antigen affinity chromatography
RUO
29735, 9194
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
O60669, Q63344
SLC16A7
Synthetic peptide of human MCT2.
100 μL
Primary
Human, Rat, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.