missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCT1/SLC16A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £488.00
Specifications
| Antigen | MCT1/SLC16A1 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18203882
|
Novus Biologicals
NBP2-58825 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653688
|
Novus Biologicals
NBP2-58825-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCT1/SLC16A1 Polyclonal specifically detects MCT1/SLC16A1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| MCT1/SLC16A1 | |
| Polyclonal | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| FLJ36745, HHF7, MCT, MCT 1, MCT1MGC44475, member 1, monocarboxylate transporter 1, Solute carrier family 16 member 1, solute carrier family 16, member 1 (monocarboxylic acid transporter 1) | |
| SLC16A1 | |
| IgG | |
| Affinity Purified |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Human | |
| 6566 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KEQKANEQKKESKEEETSIDVAGKPNEVTKAAESPDQKDTDGGPKEEESPV | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title