missing translation for 'onlineSavingsMsg'
Learn More

MCT1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen MCT1
Concentration 0.29 mg/mL
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16899634
missing translation for 'viewDocuments'
Proteintech
20139-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
16889634
missing translation for 'viewDocuments'
Proteintech
20139-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SLC16A1 is a monocarboxylate transporter (MCT1) that mediates the movement of lactate and pyruvate across cell membranes. Import and export of these substrates by tissues such as erythrocytes, muscle, intestine, and kidney are ascribed largely to the action of a proton-coupled MCT.
Specifications

Specifications

MCT1
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunoprecipitation
Unconjugated
Rabbit
Human
P53985
6566
MCT1 Fusion Protein Ag14098
Primary
-20°C
SLC16A1
0.29 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
MCT, MCT 1, MCT1, Monocarboxylate transporter 1, SLC16A1
SLC16A1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.