missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA576687
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
SLC16A1 is a monocarboxylate transporter (MCT1) that mediates the movement of lactate and pyruvate across cell membranes. Import and export of these substrates by tissues such as erythrocytes, muscle, intestine, and kidney are ascribed largely to the action of a proton-coupled MCT.
Specifications
| MCT1 | |
| Polyclonal | |
| Unconjugated | |
| SLC16A1 | |
| AL022710; FLJ36745; HHF7; MCT; MCT 1; Mct1; MCT1D; MGC44475; monocarboxylate transporter; Monocarboxylate transporter 1; RATMCT1; RNMCT1; SLC16A1; SLC16A1 protein; solute carrier 16 (monocarboxylic acid transporter), member 1; solute carrier family 16 (monocarboxylate transporter), member 1; solute carrier family 16 (monocarboxylic acid transporters), member 1; solute carrier family 16 member 1; solute carrier family 16, member 1 (monocarboxylic acid transporter 1) | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20501, 25027, 6566 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P53985, P53986, P53987 | |
| SLC16A1 | |
| Recombinant full length Human MCT1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction