missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCRS1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558639
This item is not returnable.
View return policy
Description
Immunogen sequence: HPDAFYLART AKALQAHWQL MKQYYLLEDQ TVQPLPKGDQ VLNFSDAEDL IDDSKLKDMR DEVLEHELMV ADRRQKREIR QLEQELHKWQ VLVD Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
Modulates the transcription repressor activity of DAXX by recruiting it to the nucleolus (PubMed:11948183). As part of the NSL complex it may be involved in acetylation of nucleosomal histone H4 on several lysine residues (PubMed:20018852). Putative regulatory component of the chromatin remodeling INO80 complex which is involved in transcriptional regulation, DNA replication and probably DNA repair. May also be an inhibitor of TERT telomerase activity (PubMed:15044100). Binds to G-quadruplex structures in mRNA (PubMed:16571602). Binds to RNA homopolymer poly(G) and poly(U) (PubMed:16571602). [UniProt].
Specifications
| MCRS1 | |
| Polyclonal | |
| Unconjugated | |
| MCRS1 | |
| 58 kDa microspherule protein; C78274; cell cycle-regulated factor (78 kDa); cell cycle-regulated factor p78; ICP22BP; INO80 complex subunit J; INO80 complex subunit Q; INO80Q; Mcrs1; MCRS2; Microspherule protein 1; MSP58; nucleolar protein; P78 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10445, 300222, 51812 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96EZ8, Q99L90 | |
| MCRS1 | |
| Recombinant protein corresponding to Human MCRS1. Recombinant protein control fragment (Product #RP-97876). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction