missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCPIP1/ZC3H12A Antibody [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-18333JF525
This item is not returnable.
View return policy
Description
MCPIP1/ZC3H12A Polyclonal antibody specifically detects MCPIP1/ZC3H12A in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| MCPIP1/ZC3H12A | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Janelia Fluor 525 | |
| Reg1, Regnase-1, zinc finger CCCH-type containing 12A | |
| Antibody was raised against a peptide corresponding to 14 amino acids near the carboxyl terminus of human MCPIP1/ZC3H12A. The immunogen is located within the last 50 amino acids of MCPIP1/ZC3H12A. | |
| 0.1 mL | |
| Cancer, Cell Biology, Editing and Processing Endonucleases, Immunology, Inflammation | |
| 80149 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction