missing translation for 'onlineSavingsMsg'
Learn More

MCPIP1/ZC3H12A Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18428747 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428747 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428747 Supplier Novus Biologicals Supplier No. NBP176258AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MCPIP1/ZC3H12A Polyclonal antibody specifically detects MCPIP1/ZC3H12A in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCPIP1/ZC3H12A
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias Reg1, Regnase-1, zinc finger CCCH-type containing 12A
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide near the center of human ZC3H12A. The immunogen is located within amino acids 330 - 380 of ZC3H12A. Amino Acid Squence: PPRAPSKDKNGRRPSPS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Editing and Processing Endonucleases, Immunology, Inflammation
Primary or Secondary Primary
Gene ID (Entrez) 80149
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.