missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCPH1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575562
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This protein is implicated in chromosome condensation and DNA damage induced cellular responses, and may play a role in neurogenesis and regulation of the size of the cerebral cortex.
Specifications
| MCPH1 | |
| Polyclonal | |
| Unconjugated | |
| MCPH1 | |
| 5430437K10Rik; BRCT-repeat inhibitor of TERT expression 1; BRIT 1; BRIT1; D030046N04Rik; FLJ12847; Mcph1; MCT; Microcephalin; microcephalin 1; microcephaly, primary autosomal recessive 1; RGD1560642; truncated microcephalin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 244329, 306594, 79648 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7TT79, Q8NEM0 | |
| MCPH1 | |
| Synthetic peptide corresponding to amino acids 141-186 of Human MCPH1. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction