missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCPH1 Antibody (OTI2A3), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72600APC
This item is not returnable.
View return policy
Description
MCPH1 Monoclonal antibody specifically detects MCPH1 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| MCPH1 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI2A3 | |
| Western Blot, Immunohistochemistry | |
| BRCT-repeat inhibitor of TERT expression 1, BRIT1, FLJ12847, MCT, microcephalin, microcephalin 1, microcephaly, primary autosomal recessive 1 | |
| Human recombinant protein fragment corresponding to amino acids 1-264 of human MCPH1 (NP_078872) produced in E.coli. | |
| 0.1 mL | |
| DNA Repair | |
| 79648 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction