missing translation for 'onlineSavingsMsg'
Learn More

MCPA-carboxylic acid, Dr. Ehrenstorfer

Product Code. 30324163
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324163 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324163 Supplier Dr. Ehrenstorfer Supplier No. DREC14763200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material MCPA-carboxylic acid
CAS 334758-22-4
Analyte or Component Names MCPA-carboxylic acid
Shipping Condition Room Temperature
Molecular Formula C9 H7 Cl O5
Certifications/Compliance ISO/IEC 17025
Synonym Benzoic acid, 2-(carboxymethoxy)-5-chloro-, 2-(Carboxymethoxy)-5-chlorobenzoic acid, NSC 525058
InChI Formula InChI=1S/C9H7ClO5/c10-5-1-2-7(15-4-8(11)12)6(3-5)9(13)14/h1-3H,4H2,(H,11,12)(H,13,14)
Solution Type Neat
SMILES OC(=O)COc1ccc(Cl)cc1C(=O)O
IUPAC Name 2-(carboxymethoxy)-5-chlorobenzoic acid
Industry Type Environmental Testing
Molecular Weight (g/mol) 230.6
Quantity 10 mg
Formula Weight 229.9982 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.