missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mcp1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | Mcp1 |
|---|---|
| Applications | Western Blot, Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17442333
|
Proteintech
26161-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17432333
|
Proteintech
26161-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Mcp1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of mouse Mcp1. This antibody recognizes human, mouse antigen. The Mcp1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| Mcp1 | |
| Polyclonal | |
| Rabbit | |
| 20296 | |
| Primary |
| Western Blot, Immunofluorescence, ELISA | |
| Unconjugated | |
| Human, Mouse | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title