missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCP1 Rabbit anti-Human, CoraLite 650, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL650-25542-100UL
This item is not returnable.
View return policy
Description
MCP1 Polyclonal antibody apecifically detetects MCP1 antigen in Human Samples. It is validated for Flow Cytometry.Specifications
| MCP1 | |
| Polyclonal | |
| CoraLite 650 | |
| BC009716 | |
| Rabbit | |
| Antigen affinity purification | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| CCL2 | |
| MCP1 fusion protein Ag10601 | |
| 100 μL | |
| 6347 | |
| Store at -20°C. Avoid exposure to light. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction