Learn More
PeproTech MCP-2 Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P35-50UG
Description
AA Sequence of recombinant protein: QPDSVSIPIT CCFNVINRKI PIQRLESYTR ITNIQCPKEA VIFKTKRGKE VCADPKERWV RDSMKHLDQI FQNLKP. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human MCP-2 (CCL8). Anti-Human MCP-2 (CCL8)-specific antibody was purified by affinity chromatography employing an immobilized Human MCP-2 (CCL8) matrix. Sandwich ELISA: To detect Human MCP-2 (CCL8) by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human MCP-2 (CCL8) (500-P35Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human MCP-2 (CCL8). Western Blot: To detect hMCP-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hMCP-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
This antimicrobial gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of N-terminal cysteine residues of the mature peptide. This chemokine is a member of the CC subfamily which is characterized by two adjacent cysteine residues. This cytokine displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils. By recruiting leukocytes to sites of inflammation this cytokine may contribute to tumor-associated leukocyte infiltration and to the antiviral state against HIV infection.
Specifications
| MCP-2 | |
| Polyclonal | |
| Unconjugated | |
| CCL8 | |
| 1810063B20Rik; AB023418; C-C motif chemokine 8; C-C motif chemokine ligand 8; CCL8; chemokine (C-C motif) ligand 8; HC14; MCP2; MCP-2; MCP-2(6-76); monocyte chemoattractant protein 2; monocyte chemoattractant protein-2; monocyte chemotactic protein 2; SCYA10; SCYA8; small inducible cytokine A8; small inducible cytokine subfamily A (Cys-Cys), member 8 (monocyte chemotactic protein 2); small-inducible cytokine A8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6355 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P80075 | |
| CCL8 | |
| E.coli-derived Recombinant Human MCP-2 (CCL8). | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.