missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCP-2 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148453
This item is not returnable.
View return policy
Description
Reconstitute at 1 mg/mL in sterile PBS.
This antimicrobial gene is one of several chemokine genes clustered on the q-arm of chromosome 17. Chemokines form a superfamily of secreted proteins involved in immunoregulatory and inflammatory processes. The superfamily is divided into four subfamilies based on the arrangement of N-terminal cysteine residues of the mature peptide. This chemokine is a member of the CC subfamily which is characterized by two adjacent cysteine residues. This cytokine displays chemotactic activity for monocytes, lymphocytes, basophils and eosinophils. By recruiting leukocytes to sites of inflammation this cytokine may contribute to tumor-associated leukocyte infiltration and to the antiviral state against HIV infection.
Specifications
| MCP-2 | |
| Polyclonal | |
| Unconjugated | |
| CCL8 | |
| 1810063B20Rik; AB023418; C-C motif chemokine 8; C-C motif chemokine ligand 8; CCL8; chemokine (C-C motif) ligand 8; HC14; MCP2; MCP-2; MCP-2(6-76); monocyte chemoattractant protein 2; monocyte chemoattractant protein-2; monocyte chemotactic protein 2; SCYA10; SCYA8; small inducible cytokine A8; small inducible cytokine subfamily A (Cys-Cys), member 8 (monocyte chemotactic protein 2); small-inducible cytokine A8 | |
| Goat | |
| Protein A/G | |
| RUO | |
| 6355 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Neutralization, Western Blot | |
| 1 mg/mL | |
| PBS with trehalose and no preservative | |
| P80075 | |
| CCL8 | |
| E. coli-derived recombinant human CCL8/MCP-2, Gln24-Pro99. | |
| 1 mg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction