missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCP-1 Recombinant Rabbit Monoclonal Antibody (123), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£132.00 - £377.00
Specifications
| Antigen | MCP-1 |
|---|---|
| Clone | 123 |
| Concentration | 100 μg/mL |
| Applications | ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30019254
![]() |
Invitrogen™
MA552140 |
100 μL |
£377.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30019715
|
Invitrogen™
MA552141 |
20 μL |
£132.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCP-1 Recombinant Monoclonal antibody specifically detects MCP-1 in Human samples. It is validated for ELISASpecifications
| MCP-1 | |
| 100 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative | |
| Acidic seminal fluid protein; AI323594; C-C motif chemokine 2; C-C motif chemokine ligand 2; Ccl2; chemokine (C-C motif) ligand 2; GDCF-2; HC11; H-MCP-1; HSMCR30; immediate-early serum-responsive JE protein; immediate-early serum-responsive protein JE; Je; MCAF; Mcp1; MCP-1; MCP1A; MCP-1A; MGC9434; Monocyte chemoattractant protein 1; monocyte chemoattractant protein-1; monocyte chemotactic and activating factor; Monocyte chemotactic protein 1; monocyte chemotactic protein 1A; Monocyte secretory protein JE; monocyte-chemoattractant protein-1 precursor; platelet-derived growth factor-inducible protein JE; RP23-350G1.3; SCYA2; Sigje; small inducible cytokine A2; small inducible cytokine A2 (monocyte chemotactic protein 1, homologous to mouse Sig-je); small inducible cytokine subfamily A (Cys-Cys), member 2; small inducible gene JE; small-inducible cytokine A2; SMC-CF | |
| Recombinant Human MCP-1/CCL2 Protein (Gln24-Thr99). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Ccl2 |
| 123 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P13500 | |
| 6347 | |
| IgG | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title