missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCMBP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-43603
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Specifically detects MCMBP in Human samples, and it is validated for Western Blotting
MCMBP Polyclonal specifically detects MCMBP in Human samples. It is validated for Western Blot.
Especificaciones
| MCMBP | |
| Polyclonal | |
| Western Blot 1:500-1:3000 | |
| C10orf119, FLJ13081, MCM-binding protein, MCM-BP, minichromosome maintenance complex binding protein, mini-chromosome maintenance complex-binding protein, minichromosome maintenance complex-binding protein | |
| Rabbit | |
| 73 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| MCMBP | |
| Recombinant protein encompassing a sequence within the center region of human MCMBP. The exact sequence is proprietary. | |
| Immunogen affinity purified | |
| RUO | |
| 79892 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido