missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCM9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549293
This item is not returnable.
View return policy
Description
Predicted to react with bovine based on sequence homology.
Component of the MCM8-MCM9 complex, a complex involved in homologous recombination repair following DNA interstrand cross-links and plays a key role during gametogenesis. The MCM8- MCM9 complex probably acts as a hexameric helicase downstream of the Fanconi anemia proteins BRCA2 and RAD51 and is required to process aberrant forks into homologous recombination substrates and to orchestrate homologous recombination with resection, fork stabilization and fork restart.
Specifications
| MCM9 | |
| Polyclonal | |
| Unconjugated | |
| MCM9 | |
| 9030408O17Rik; C6orf61; dJ329L24.1; dJ329L24.3; DNA helicase MCM9; DNA replication licensing factor MCM9; Gm235; hMCM9; MCM9; Mcmdc1; Minichromosome maintenance 9; minichromosome maintenance 9 homologous recombination repair factor; minichromosome maintenance complex component 9; mini-chromosome maintenance deficient 9; minichromosome maintenance deficient domain containing 1; mini-chromosome maintenance deficient domain-containing protein 1; ODG4; RGD1560557 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 254394 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9NXL9 | |
| MCM9 | |
| KLH conjugated synthetic peptide between 316-345 amino acids from the C-terminal region of human MCM9. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction