Learn More
MCM9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556653
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SPPPERKNRG ERGPSSPPTT TAPMRVSKRK SFQLRGSTEK LIVSKESLFT LPELGDEAFD CDWD Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 53%.
Component of the MCM8-MCM9 complex, a complex involved in homologous recombination repair following DNA interstrand cross-links and plays a key role during gametogenesis. The MCM8- MCM9 complex probably acts as a hexameric helicase downstream of the Fanconi anemia proteins BRCA2 and RAD51 and is required to process aberrant forks into homologous recombination substrates and to orchestrate homologous recombination with resection, fork stabilization and fork restart.Specifications
MCM9 | |
Polyclonal | |
Unconjugated | |
MCM9 | |
9030408O17Rik; C6orf61; dJ329L24.1; dJ329L24.3; DNA helicase MCM9; DNA replication licensing factor MCM9; Gm235; hMCM9; MCM9; Mcmdc1; Minichromosome maintenance 9; minichromosome maintenance 9 homologous recombination repair factor; minichromosome maintenance complex component 9; mini-chromosome maintenance deficient 9; minichromosome maintenance deficient domain containing 1; mini-chromosome maintenance deficient domain-containing protein 1; ODG4; RGD1560557 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
254394 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NXL9 | |
MCM9 | |
Recombinant protein corresponding to Human MCM9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only