missing translation for 'onlineSavingsMsg'
Learn More

MCM9 Antibody, Novus Biologicals™

Product Code. 18409509 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18409509 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18409509 Supplier Novus Biologicals Supplier No. H00254394D01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MCM9 Polyclonal antibody specifically detects MCM9 in Human, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM9
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_694987.1
Gene Alias C6orf61, chromosome 6 open reading frame 61, dJ329L24.1, dJ329L24.3, DNA replication licensing factor MCM9, FLJ13942, FLJ20170, FLJ56845, hMCM9, MCMDC1, MGC35304, minichromosome maintenance complex component 9, Mini-chromosome maintenance deficient 9, minichromosome maintenance deficient domain containing 1, Mini-chromosome maintenance deficient domain-containing protein 1
Host Species Rabbit
Immunogen MCM9 (NP_694987.1, 1 a.a. - 391 a.a.) full-length human protein. MNSDQVTLVGQVFESYVSEYHKNDILLILKERDEDAHYPVVVNAMTLFETNMEIGEYFNMFPSEVLTIFDSALRRSALTILQSLSQPEAVSMKQNLHARISGLPVCPELVREHIPKTKDVGHFLSVTGTVIRTSLVKVLEFERDYMCNKCKHVFVIKADFEQYYTFCRPSSCPSLESCDSSKFTCLSGLSSSPTRCRDYQEIKIQEQVQRLSVGSIPRSMKVILEDDLVDSCKSGDDLTIYGIVMQRWKPFQQDVRCEVEIVLKANYIQVNNEQSSGIIMDEEVQKEFEDFWEYYKSDPFAGRNVILASLCPQVFGMYLVKLAVAMVLAGGIQRTDATGTRVRGESHLLLVGDPGTGKSQFLKYAAKITPRSVLTTGIGSTSAGIVCDNFK
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254394
Target Species Human, Rat
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.