missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM8 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£162.00 - £375.00
Specifications
| Antigen | MCM8 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18319751
|
Novus Biologicals
NBP3-05116-20UL |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18317203
|
Novus Biologicals
NBP3-05116-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCM8 Polyclonal specifically detects MCM8 in Human samples. It is validated for Western Blot.Specifications
| MCM8 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| C20orf154, chromosome 20 open reading frame 154, dJ967N21.5, DNA replication licensing factor MCM8, MCM8 minichromosome maintenance deficient 8, MGC119522, MGC119523, MGC12866, MGC4816, Minichromosome maintenance 8, minichromosome maintenance complex component 8, REC, REC homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 591-840 of human MCM8 (NP_115874.3). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA Repair | |
| PBS with 50% glycerol, pH7.3. | |
| 84515 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title