missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM8 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Description
MCM8 Polyclonal antibody specifically detects MCM8 in Human samples. It is validated for Immunofluorescence
Specifications
Specifications
| Antigen | MCM8 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Formulation | PBS, pH 7.2, 40% glycerol |
| Gene Alias | C20orf154, chromosome 20 open reading frame 154, dJ967N21.5, DNA replication licensing factor MCM8, MCM8 minichromosome maintenance deficient 8, MGC119522, MGC119523, MGC12866, MGC4816, Minichromosome maintenance 8, minichromosome maintenance complex component 8, REC, REC homolog |
| Host Species | Rabbit |
| Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids: GDTVTITGIVKVSNAEEGSRNKNDKCMFLLYIEANSISNSKGQKTKSSEDGCKHGMLMEFSLKDLYAIQEIQAEENLFKLIVNSLCPVIFGHELVKA |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?