Learn More
MCM8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA565399
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: ERKGSILVDFK ELTEGGEVTN LIPDIATELR DAPEKTLACM GLAIHQVLTK DLERHAAELQ AQEGLSNDGE TMVNVPHIHA RVYNYEPLTQ LKNVRANYYG KYIALRGTVV RVSNIKPLCT KMAFLCAACG EIQSFPLPDG KYSL Highest antigen sequence identity to the following orthologs - mouse 88%, rat 87%.
The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein contains the central domain that is conserved among the MCM proteins. This protein has been shown to co-immunoprecipitate with MCM4, 6 and 7, which suggests that it may interact with other MCM proteins and play a role in DNA replication. Alternatively spliced transcript variants encoding distinct isoforms have been described.Specifications
MCM8 | |
Polyclonal | |
Unconjugated | |
MCM8 | |
5730432L01Rik; C20orf154; dJ967N21.5; DNA helicase MCM8; DNA replication licensing factor MCM8; MCM8; MCM8 minichromosome maintenance deficient 8; MGC119522; MGC119523; MGC12866; MGC4816; minichromosome maintenance 8; minichromosome maintenance 8 homologous recombination repair factor; minichromosome maintenance complex component 8; minichromosome maintenance deficient 8; minichromosome maintenance deficient 8 (S. cerevisiae); POF10; REC; REC homolog | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84515 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UJA3 | |
MCM8 | |
Recombinant Human MCM8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only