missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99005-100ul
This item is not returnable.
View return policy
Description
MCM8 Polyclonal antibody specifically detects MCM8 in Human samples. It is validated for Western Blot
Specifications
| MCM8 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C20orf154, chromosome 20 open reading frame 154, dJ967N21.5, DNA replication licensing factor MCM8, MCM8 minichromosome maintenance deficient 8, MGC119522, MGC119523, MGC12866, MGC4816, Minichromosome maintenance 8, minichromosome maintenance complex component 8, REC, REC homolog | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MCM8. | |
| 100 μg | |
| DNA Repair | |
| 84515 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction