missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | MCM8 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
MCM8 Polyclonal specifically detects MCM8 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MCM8 | |
| Polyclonal | |
| Rabbit | |
| DNA Repair | |
| C20orf154, chromosome 20 open reading frame 154, dJ967N21.5, DNA replication licensing factor MCM8, MCM8 minichromosome maintenance deficient 8, MGC119522, MGC119523, MGC12866, MGC4816, Minichromosome maintenance 8, minichromosome maintenance complex component 8, REC, REC homolog | |
| MCM8 | |
| IgG | |
| 94 kDa |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Q9UJA3 | |
| 84515 | |
| Synthetic peptides corresponding to MCM8(minichromosome maintenance complex component 8) The peptide sequence was selected from the C terminal of human MCM8. Peptide sequence IRLTEARARLELREEATKEDAEDIVEIMKYSMLGTYSDEFGNLDFERSQH. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title