missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MCM7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16389084
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16389084 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16389084 Supplier Invitrogen™ Supplier No. PA582122

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: QSPTFMPLIM CPSQECQTNR SGGRLYLQTR GSRFIKFQEM KMQEHSDQVP VGNIPRSITV LVEGENTRIA QPGDHVSVTG IFLPILRTGF RQVVQGLLSE TYLEAHRIVK MNKSEDDESG AGE Highest antigen sequence indentity to the following orthologs: Rat - 91%, Mouse - 89%.

MCM7 encodes a protein that is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 6 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme.The protein encoded by this gene is one of the highly conserved mini-chromosome maintenance proteins (MCM) that are essential for the initiation of eukaryotic genome replication. The hexameric protein complex formed by the MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. The MCM complex consisting of this protein and MCM2, 4 and 6 proteins possesses DNA helicase activity, and may act as a DNA unwinding enzyme. Cyclin D1-dependent kinase, CDK4, is found to associate with this protein, and may regulate the binding of this protein with the tumorsuppressor protein RB1/RB. Alternatively spliced transcript variants encoding distinct isoforms have been reported.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Mcm7
Gene Accession No. P33993, Q61881
Gene Alias AI747533; CDC47; CDC47 homolog; CDC47 homolog A; CDC47p; DNA replication licensing factor MCM7; DNA replication licensing factor mcm7-A; DNA replication licensing factor MCM7-like protein; homolog of S. cerevisiae Cdc47; mCDC47; MCM2; MCM7; MCM7 minichromosome maintenance deficient 7; mcm7.S; mcm7-a; mcm7-b; Mcmd7; mini chromosome maintenance deficient 7; minichromosome maintenance complex component 7; minichromosome maintenance complex component 7 S homeolog; minichromosome maintenance deficient 7; minichromosome maintenance deficient 7 (S. cerevisiae); minichromosome maintenance protein 7; minichromosome maintenance protein 7-A; P1 P85MCM; P1.1-MCM3; P1CDC47; P85MCM; p90; PNAS146; PPP1R104; protein phosphatase 1, regulatory subunit 104; Unknown (protein for MGC:128845); XELAEV_18000877mg; XMCM7; xMCM7-A
Gene Symbols Mcm7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MCM7. Recombinant protein control fragment (Product #RP-102268).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17220, 288532, 4176
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.