missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MCM7 Monoclonal specifically detects MCM7 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.
Specifications
Specifications
| Antigen | MCM7 |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
| Classification | Monoclonal |
| Clone | rMCM7/1468 |
| Concentration | 0.2 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Alias | CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?