missing translation for 'onlineSavingsMsg'
Learn More

MCM7 Mouse anti-Human, Clone: rMCM7/1468, Novus Biologicals™

Mouse Monoclonal Antibody

£387.00

Specifications

Antigen MCM7
Clone rMCM7/1468
Concentration 0.2 mg/mL
Dilution Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array
Applications Western Blot, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18601688
View Documents
Novus Biologicals
NBP3-07277-100ug
100 μg
£387.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MCM7 Monoclonal specifically detects MCM7 in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

MCM7
0.2 mg/mL
Western Blot, Immunohistochemistry (Paraffin), Peptide Array
Unconjugated
Mouse
Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, Stem Cell Markers
10 mM PBS with 0.05% BSA
4176
IgG1 κ
Protein A or G purified
rMCM7/1468
Western Blot 1 to 2 μg/mL, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array
Monoclonal
Purified
RUO
Canine, Human
CDC47 homolog, CDC47P85MCM, DNA replication licensing factor MCM7, EC 3.6.4.12, homolog of S. cerevisiae Cdc47, MCM2PNAS146, MCM7 minichromosome maintenance deficient 7 (S. cerevisiae), minichromosome maintenance complex component 7, minichromosome maintenance deficient (S. cerevisiae) 7, minichromosome maintenance deficient 7, P1.1-MCM3, P1CDC47
Recombinant human MCM7 protein fragment (aa195-319) (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.