missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCM6 Recombinant Rabbit Monoclonal Antibody (23GB2005), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553194
This item is not returnable.
View return policy
Description
MCM6 may be involved in the control of a single round of DNA replication during S phase. It binds to chromatin during G1 and detaches from it during S phase as if it licenses the chromatin to replicate.
Specifications
| MCM6 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Mcm6 | |
| ASP-l1; D1Wsu22e; DNA replication licensing factor MCM6; intestinal DNA replication protein; MCG40308; Mcm6; MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe); Mcmd6; mini chromosome maintenance deficient 6; minichromosome maintenance complex component 6; minichromosome maintenance deficient (mis5, S. pombe) 6; minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe); minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S. cerevisiae); Mis5; Mis5 homolog; p105MCM | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 17219, 29685, 4175 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2005 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P97311, Q14566, Q62724 | |
| Mcm6 | |
| A synthesized peptide derived from human MCM6 (601-821AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction