missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM6 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33270-100ul
This item is not returnable.
View return policy
Description
MCM6 Monoclonal antibody specifically detects MCM6 in Human,Mouse samples. It is validated for ELISA,Western Blot
Specifications
| MCM6 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| DNA replication licensing factor MCM6, EC 3.6.4.12, MCG40308, MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe), MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S.cerevisiae), minichromosome maintenance complex component 6, minichromosome maintenance deficient (mis5, S. pombe) 6, minichromosome maintenance deficient 6 homolog, minichromosome maintenance deficient 6 homolog (S. cerevisiae), Mis5, MIS5 homolog, p105MCM | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 601-821 of human MCM6 (NP_005906.2).,, Sequence:, LRQRDGSGVTKSSWRITVRQLESMIRLSEAMARMHCCDEVQPKHVKEAFRLLNKSIIRVETPDVNLDQEEEIQMEVDEGAGGINGHADSPAPVNGINGYNEDINQESAPKASLRLGFSEYCRISNLIVLHLRKVEEEEDESALKRSELVNWYLKEIESEIDSEEELINKKRIIEKVIHRLTHYDHVLIELTQAGLKGSTEGSESYEEDPYLVVNPNYLLED | |
| 100 μL | |
| Cell Cycle and Replication, DNA Repair | |
| 4175 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction