missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM6 Antibody (MCM6/2999), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-79862CL1
This item is not returnable.
View return policy
Description
MCM6 Monoclonal antibody specifically detects MCM6 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Protein Array
Specifications
| MCM6 | |
| Monoclonal | |
| CoraFluor 1 | |
| DNA replication licensing factor MCM6, EC 3.6.4.12, MCG40308, MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe), MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S.cerevisiae), minichromosome maintenance complex component 6, minichromosome maintenance deficient (mis5, S. pombe) 6, minichromosome maintenance deficient 6 homolog, minichromosome maintenance deficient 6 homolog (S. cerevisiae), Mis5, MIS5 homolog, p105MCM | |
| Recombinant fragment (around aa 228-368) of human MCM6 protein (exact sequence is proprietary) (Uniprot: Q14566) | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair | |
| 4175 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| MCM6/2999 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction