missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
MCM6 Monoclonal antibody specifically detects MCM6 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
Specifications
Specifications
| Antigen | MCM6 |
| Applications | Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA |
| Classification | Monoclonal |
| Clone | 7D8 |
| Conjugate | Unconjugated |
| Formulation | In 1x PBS, pH 7.4 |
| Gene Accession No. | NP_005906.2 |
| Gene Alias | DNA replication licensing factor MCM6, EC 3.6.4.12, MCG40308, MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe), MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S.cerevisiae), minichromosome maintenance complex component 6, minichromosome maintenance deficient (mis5, S. pombe) 6, minichromosome maintenance deficient 6 homolog, minichromosome maintenance deficient 6 homolog (S. cerevisiae), Mis5, MIS5 homolog, p105MCM |
| Host Species | Mouse |
| Immunogen | MCM6 (NP_005906.2, 1 a.a. ~ 821 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MDLAAAAEPGAGSQHLEVRDEVAEKCQKLFLDFLEEFQSSDGEIKYLQLAEELIRPERNTLVVSFVDLEQFNQQLSTTIQEEFYRVYPYLCRALKTFVKDRKEIPLAKDFYVAFQDLPTRHKIRELTSSRIGLLTRISGQVVRTHPVHPELVSGTFLCLDCQTVIRDVEQQFKYTQPNICRNPVCANRRRFLLDTNKSRFVDFQKVRIQETQAELPRGSIPRSLEVILRAEAVESAQAGDKCDFTGTLIVVPDVSKLSTPGARAETNSRVSGVDGYETEGIRGLRALGVRDLSYRLVFLACCVAPTNPRFGGKELRDEEQTAESIKNQMTVKEWEKVFEMSQDKNLYHNLCTSLFPTIHGNDEVKRGVLLMLFGGVPKTTGEGTSLRGDINVCIVGDPSTAKSQFLKHVEEFSPRAVYTSGKASSAAGLTAAVVRDEESHEFVIEAGALMLADNGVCCIDEFDKMDVRDQVAIHEAMEQQTISITKAGVKATLNARTSILAAANPISGHYDRSKSLKQNINLSAPIMSRFDLFFILVDECNEVTDYAIARRIVDLHSRIEESIDRVYSLDDIRRYLLFARQFKPKISKESEDFIVEQYKHLRQRDGSGVTKSSWRITVRQLESMIRLSEAMARMHCCDEVQPKHVKEAFRLLNKSIIRVETPDVNLDQEEEIQMEVDEGAGGINGHADSPAPVNGINGYNEDINQESAPKASLRLGFSEYCRISNLIVLHLRKVEEEEDESALKRSELVNWYLKEIESEIDSEEELINKKRIIEKVIHRLTHYDHVLIELTQAGLKGSTEGSESYEEDPYLVVNPNYLLED |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?