missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM4 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-290F
This item is not returnable.
View return policy
Description
MCM4 Polyclonal antibody specifically detects MCM4 in Human, Mouse samples. It is validated for Immunohistochemistry
Specifications
| MCM4 | |
| Polyclonal | |
| Immunohistochemistry | |
| CDC21, CDC21 homolog, CDC54, DNA replication licensing factor MCM4, EC 3.6.4.12, hCdc21, homolog of S. pombe cell devision cycle 21, MCM4 minichromosome maintenance deficient 4, MCM4 minichromosome maintenance deficient 4 (S. cerevisiae), MGC33310, minichromosome maintenance complex component 4, minichromosome maintenance deficient 4, P1-Cdc21 | |
| The immunogen recognized by this antibody maps to a region between residues 750 and 800 of human Minichromosome Maintenance 4 using the numbering given in entry NP_005905.2 (GeneID 4173). | |
| 0.1 mL | |
| Cell Cycle and Replication, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Stem Cell Markers | |
| 4173 | |
| Store at 4C in the dark. | |
| IgG |
| Immunohistochemistry | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction