missing translation for 'onlineSavingsMsg'
Learn More

MCM4 Antibody (1A6), Novus Biologicals™

Product Code. 18350038 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18350038 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18350038 Supplier Novus Biologicals Supplier No. H00004173M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MCM4 Monoclonal antibody specifically detects MCM4 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM4
Applications ELISA
Classification Monoclonal
Clone 1A6
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH31061
Gene Alias CDC21, CDC21 homolog, CDC54, DNA replication licensing factor MCM4, EC 3.6.4.12, hCdc21, homolog of S. pombe cell devision cycle 21, MCM4 minichromosome maintenance deficient 4, MCM4 minichromosome maintenance deficient 4 (S. cerevisiae), MGC33310, minichromosome maintenance complex component 4, minichromosome maintenance deficient 4, P1-Cdc21
Host Species Mouse
Immunogen MCM4 (AAH31061, 267 a.a. ~ 366 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. KTKNMRNLNPEDIDQLITISGMVIRTSQLIPEMQEAFFQCQVCAHTTRVEMDRGRIAEPSVCGRCHTTHSMALIHNRSLFSDKQMIKLQESPEDMPAGQT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Core ESC Like Genes, DNA Double Strand Break Repair, DNA Repair, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4173
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.