missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM3AP Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-71882PE
This item is not returnable.
View return policy
Description
MCM3AP Polyclonal antibody specifically detects MCM3AP in Human samples. It is validated for Western Blot, Immunoprecipitation (Negative)
Specifications
| MCM3AP | |
| Polyclonal | |
| Western Blot, Immunoprecipitation (Negative) | |
| 80 kDa MCM3-associated protein, FLJ44336, GANPFLJ45306, human mRNA for MCM3 import factor10Protein GANP, KIAA0572MAP80germinal center-associated nuclear protein, Map80, MCM3 import protein, MCM3 minichromosome maintenance deficient 3 (S. cerevisiae) associated protein, MCM3 minichromosome maintenance deficient 3 associated protein, minichromosome maintenance complex component 3 associated protein, minichromosome maintenance deficient (S. cerevisiae) 3-associated protein, minichromosome maintenance deficient 3-associated protein | |
| The immunogen for this product maps to a region between residue 1 and 50 of human Germinal center-Associated Nuclear Protein using the numbering given in entry NP_003897.2 (GeneID 8888). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 8888 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction