missing translation for 'onlineSavingsMsg'
Learn More

MCM3AP Antibody, DyLight 755, Novus Biologicals™

Product Code. 18420925 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420925 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420925 Supplier Novus Biologicals Supplier No. NBP171882IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MCM3AP Polyclonal antibody specifically detects MCM3AP in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM3AP
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias 80 kDa MCM3-associated protein, FLJ44336, GANPFLJ45306, human mRNA for MCM3 import factor10Protein GANP, KIAA0572MAP80germinal center-associated nuclear protein, Map80, MCM3 import protein, MCM3 minichromosome maintenance deficient 3 (S. cerevisiae) associated protein, MCM3 minichromosome maintenance deficient 3 associated protein, minichromosome maintenance complex component 3 associated protein, minichromosome maintenance deficient (S. cerevisiae) 3-associated protein, minichromosome maintenance deficient 3-associated protein
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1 and 50 of human Germinal center-Associated Nuclear Protein using the numbering given in entry NP_003897.2 (GeneID 8888).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8888
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.