missing translation for 'onlineSavingsMsg'
Learn More

MCM2 Rabbit anti-Human, Mouse, Rat, Clone: 0Q10S9, Novus Biologicals™

Product Code. 18377731 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377731 20 μg 20µL
18325172 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18377731 Supplier Bio-Techne Supplier No. NBP31606620UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

MCM2 Monoclonal antibody specifically detects MCM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen MCM2
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 0Q10S9
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:100 - 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias BM28MGC10606, CCNL 1, CCNL1, CDCL1mitotin, EC 3.6.4.12, KIAA0030MITOTIN, MCM2 minichromosome maintenance deficient 2, mitotin (S. cerevisiae), minichromosome maintenance complex component 2, minichromosome maintenance deficient (S. cerevisiae) 2 (mitotin), minichromosome maintenance deficient 2 (mitotin)
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MCM2 (P49736).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 4171
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.