Learn More
Invitrogen™ MCM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579645
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Testis Tissue, Rat Kidney Tissue, Mouse Intestine Tissue, Mouse Testis Tissue, Mouse Ovary Tissue, Mouse Kidney Tissue. IHC: Rat Intestinal Lymphocyte Tissue, Mouse Intestine Tissue. ICC/IF: HEPA1-6 cell, HEPA Cell, NIH3T3 Cell. Flow: Neuro-2a cell.
MCM2 (Mini chromosome maintenance protein 2) is involved in regulating DNA replication. MCM2 (also called CDCL1, mitotin and BM28), is a human nuclear protein that is crucial in the cell cycle, being involved in the onset of DNA replication and cell division. It is similar to members of the family of early S-phase proteins. MCM2 has been mapped to 3q21. From its localization, CDCL1 became a candidate for an oncogene affected by chromosomal breaks in acute myeloid leukemia (AML).
Specifications
| MCM2 | |
| Polyclonal | |
| Unconjugated | |
| MCM2 | |
| AA959861; AW476101; BM28; CCNL1; cdc19; CDCL1; cell devision cycle-like 1; cyclin-like 1; D3S3194; DFNA70; DNA replication licensing factor MCM2; Kiaa0030; MCM DNA helicase complex subunit MCM2; Mcm2; Mcmd2; MGC10606; mini chromosome maintenance deficient 2; minichromosome maintenance complex component 2; minichromosome maintenance deficient 2 (mitotin); minichromosome maintenance deficient 2 mitotin; minichromosome maintenance deficient 2 mitotin (S. cerevisiae); Minichromosome maintenance protein 2; Minichromosome maintenance protein 2 homolog; MITOTIN; mKIAA0030; Nuclear protein BM28; YBL023C; YBL0438 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 17216, 312538, 4171 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| P49736, P97310 | |
| MCM2 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of mouse MCM2 (884-904aa DLFKFNKFSRDLKRKLILQQF). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.