missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM2 Antibody (MCM2/8006R), Novus Biologicals™
Rabbit Monoclonal Antibody
£224.00 - £412.00
Specifications
| Antigen | MCM2 |
|---|---|
| Clone | MCM2/8006R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18650344
|
Novus Biologicals
NBP3-20405-100ug |
100 μg |
£412.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18622964
|
Novus Biologicals
NBP3-20405-20ug |
20 μg |
£224.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCM2 Monoclonal antibody specifically detects MCM2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| MCM2 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Core ESC Like Genes, DNA Repair, DNA replication Transcription Translation and Splicing, Phospho Specific, Stem Cell Markers | |
| 10mM PBS with 0.05% BSA | |
| 4171 | |
| IgG κ | |
| Protein A or G purified |
| MCM2/8006R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BM28MGC10606, CCNL 1, CCNL1, CDCL1mitotin, EC 3.6.4.12, KIAA0030MITOTIN, MCM2 minichromosome maintenance deficient 2, mitotin (S. cerevisiae), minichromosome maintenance complex component 2, minichromosome maintenance deficient (S. cerevisiae) 2 (mitotin), minichromosome maintenance deficient 2 (mitotin) | |
| Recombinant fragment (aa 1-200) of human MCM2 protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title