missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£154.00 - £434.00
Specifications
| Antigen | MCM10 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30229825
|
Novus Biologicals
NBP3-38397-100ul |
100 μL |
£434.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30227686
|
Novus Biologicals
NBP3-38397-20ul |
20 μL |
£154.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCM10 Polyclonal antibody specifically detects MCM10 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| MCM10 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| DNA Repair | |
| PBS (pH 7.3), 50% glycerol | |
| 55388 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| CNA43homolog of yeast MCM10, DNA43, hsMCM10, MCM10 minichromosome maintenance deficient 10, MCM10 minichromosome maintenance deficient 10 (S. cerevisiae), MGC126776, minichromosome maintenance complex component 10, PRO2249, protein MCM10 homolog | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 616-875 of human MCM10 (NP_877428.1).,, Sequence:, TNPNSIKKKQKDPQDILEVKERVEKNTMFSSQAEDELEPARKKRREQLAYLESEEFQKILKAKSKHTGILKEAEAEMQERYFEPLVKKEQMEEKMRNIREVKCRVVTCKTCAYTHFKLLETCVSEQHEYHWHDGVKRFFKCPCGNRSISLDRLPNKHCSNCGLYKWERDGMLKEKTGPKIGGETLLPRGEEHAKFLNSLK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title