missing translation for 'onlineSavingsMsg'
Learn More

Mcl1-IN-1, MedChemExpress

Product Code. 16497136
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497136 100 mg 100mg
16429657 5 mg 5mg
16427118 10 mg 10mg
16403157 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16497136 Supplier Medchem Express Supplier No. HY16669
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Mcl1-IN-1 is an inhibitor of myeloid cell factor 1 (Mcl-1) (IC50=2.4 μM).

Mcl1-IN-1 is an inhibitor of myeloid cell factor 1 (Mcl-1) (IC50=2.4 μM).

Specifications

Chemical Name or Material Mcl1-IN-1
CAS 713492-66-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H18ClN3O4
Quantity 100 mg
Solubility Information DMSO : ≥ 50 mg/mL (114.72 mM)
SMILES O=C(O)COC1=CC=CC=C1C(NC2=NC=CC=C2)C3=C(O)C4=NC=CC=C4C(Cl)=C3
Molecular Weight (g/mol) 435.86
Formula Weight 435.86
Percent Purity 98.4%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.