Learn More
Ansell Edmont™ Microgard™ 2500 Standard Protective Oversleeves
Product Code. 15576554 Shop All Ansell Products
Product Code. 15576554 Supplier Ansell Edmont™ Supplier No. WH25B0060000
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Fits over the arm just past the elbow to the bicep. Ansell Edmont™ Microgard™ 2500 Standard Protective Oversleeves include elasticated openings at both ends.
  • Elasticated at both ends
  • Bound seams
  • Microporous polypropylene laminate
TRUSTED_SUSTAINABILITY
Specifications
Size Universal
Product Type General Purpose Sleeve
Material Microporous Polypropylene Laminate
Color White
Closure Type Elastic Ends
Seam Type Bound
Cuff Style Elastic