missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCHR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11656-2-AP-150UL
This item is not returnable.
View return policy
Description
MCHR1 is a member of the G protein-coupled receptor family 1. It is an integral plasma membrane protein which binds melanin-concentrating hormone. The encoded protein can inhibit cAMP accumulation and stimulate intracellular calcium flux, and is probably involved in the neuronal regulation of food consumption. Although structurally similar to somatostatin receptors, this protein does not seem to bind somatostatin.Specifications
| MCHR1 | |
| Polyclonal | |
| Unconjugated | |
| MCHR1 | |
| G protein coupled receptor 24, GPR24, MCH 1R, MCH R1, MCH receptor 1, MCH1R, MCHR, MCHR 1, MCHR1, SLC 1, SLC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 207911, 2847, 83567 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P97639, Q8JZL2, Q99705 | |
| MCHR1 | |
| MCHR1 Fusion Protein Ag2224 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction