missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCHR1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533931
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gibbon, Monkey (100%) Mouse (85%).
MCHR1 is a member of the G protein-coupled receptor family 1. It is an integral plasma membrane protein which binds melanin-concentrating hormone. The encoded protein can inhibit cAMP accumulation and stimulate intracellular calcium flux, and is probably involved in the neuronal regulation of food consumption. Although structurally similar to somatostatin receptors, this protein does not seem to bind somatostatin.
Specifications
| MCHR1 | |
| Polyclonal | |
| Unconjugated | |
| Mchr1 | |
| AW049955; G protein coupled receptor 24; G protein-coupled receptor 24; GPCR24; GPR24; Gpr24-9 protein coupled receptor; G-protein coupled receptor 24; MCH 1 receptor; MCH receptor 1; MCH1; Mch1r; MCH-1R; MCHR; MCHR1; MCH-R1; MCHR-1; melanin concentrating hormone receptor 1; melanin-concentrating hormone receptor 1; SLC1; SLC-1; Somatostatin receptor-like protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2847 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q99705 | |
| Mchr1 | |
| Synthetic 20 amino acid peptide from N-terminal extracellular domain of human MCHR1. | |
| 50 μg | |
| Primary | |
| Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction