missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ mCherry Polyclonal Antibody
GREENER_CHOICE

Chicken Polyclonal Antibody

Brand:  Invitrogen™ PA5143576

 View more versions of this product

Product Code. 17876837

  • £430.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

mCherry Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

MCherry is a fluorophore (a fluorescent molecule) used in biotechnology as a tracer to follow the flow of fluids, as a marker when tagged to molecules and cells components. mCherry is a monomeric fluorescent construct which absorbs at 587 nm and emits at 610 nm. It is resistant to photobleaching and is stable. It matures quickly, with a t 0. 5 of 15 minutes, allowing it to be visualized soon after translation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

mCherry
Polyclonal
Unconjugated
mCherry
Full length recombinant protein expressed in and purified from E. coli.
RUO
Chemical
Antibody
IgY
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Conc. Not Determined
PBS with 0.02% sodium azide
Chicken
100 μL
Primary
4°C
Liquid
Chemical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.